missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-2-Methylbutyl [1-(5-Deoxy-beta-D-ribofuranosyl)-5-fluoro-2-oxo-1,2-dihydropyrimidin-4-yl]carbamate, Mikromol™

Product Code. 17989609
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17989609 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17989609 Supplier Mikromol Supplier No. MM0992.080025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-2-Methylbutyl [1-(5-Deoxy-beta-D-ribofuranosyl)-5-fluoro-2-oxo-1,2-dihydropyrimidin-4-yl]carbamate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5'-Deoxy-5-fluoro-N-[(2-methylbutoxy)carbonyl]cytidine
CAS 910129-15-6
Analyte or Component Names 5'-Deoxy-5-fluoro-N-[(2-methylbutoxy)carbonyl]cytidine
Shipping Condition Room Temperature
Molecular Formula C15 H22 F N3 O6
Certifications/Compliance ISO 9001
Synonym 5'-Deoxy-5-fluoro-N-[(2-methylbutoxy)carbonyl]cytidine, 2-Methylbutyl N-[1-[(2R,3R,4S,5R)-3,4-Dihydroxy-5-methyl-tetrahydrofuran-2-yl]-5-fluoro-2-oxo-pyrimidin-4-yl]carbamate, Capecitabine Imp. D (EP), (2RS)-2-Methylbutyl [1-(5-Deoxy-beta-D-ribofuranosyl)-5-fluoro-2-oxo-1,2-dihydropyrimidin-4-yl]carbamate, Capecitabine Impurity D
InChI Formula InChI=1S/C15H22FN3O6/c1-4-7(2)6-24-15(23)18-12-9(16)5-19(14(22)17-12)13-11(21)10(20)8(3)25-13/h5,7-8,10-11,13,20-21H,4,6H2,1-3H3,(H,17,18,22,23)/t7?,8-,10-,11-,13-/m1/s1
SMILES CCC(C)COC(=O)NC1=NC(=O)N(C=C1F)[C@@H]2O[C@H](C)[C@@H](O)[C@H]2O
IUPAC Name 2-methylbutyl N-[1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-2-oxopyrimidin-4-yl]carbamate
Molecular Weight (g/mol) 359.35
Quantity 25 mg
Formula Weight 359.1493 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.