missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2RS)-3-(Naphthalen-1-yloxy)propane-1,2-diol (Diol Derivative), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(2RS)-3-(Naphthalen-1-yloxy)propane-1,2-diol (Diol Derivative)
Specifications
Specifications
| Chemical Name or Material | (2RS)-3-(Naphthalen-1-yloxy)propane-1,2-diol |
| CAS | 36112-95-5 |
| Analyte or Component Names | (2RS)-3-(Naphthalen-1-yloxy)propane-1,2-diol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H14 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Propranolol Hydrochloride Imp. A (EP), Propranolol Imp. A (EP), (2RS)-3-(Naphthalen-1-yloxy)propane-1,2-diol, Diol Derivative of Propranolol, Propranolol Hydrochloride Impurity A, Propranolol Impurity A |
| InChI Formula | InChI=1S/C13H14O3/c14-8-11(15)9-16-13-7-3-5-10-4-1-2-6-12(10)13/h1-7,11,14-15H,8-9H2 |
| SMILES | OCC(O)COc1cccc2ccccc12 |
| IUPAC Name | 3-naphthalen-1-yloxypropane-1,2-diol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction