missing translation for 'onlineSavingsMsg'
Learn More

(2RS,4S)-2-[[[(2R)-2-Amino-2-phenylacetyl]amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Penilloic Acids of Ampicillin), Mikromol™

Product Code. 30051202
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30051202

Brand: Mikromol MM0253.060025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS,4S)-2-[[[(2R)-2-Amino-2-phenylacetyl]amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Penilloic Acids of Ampicillin)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Penilloic Acids of Ampicillin
CAS 124774-48-7 and 124774-49-8
Analyte or Component Names Penilloic Acids of Ampicillin
Shipping Condition Ice pack (-20°C)
Molecular Formula C15 H21 N3 O3 S
Certifications/Compliance ISO 9001
Synonym Ampicillin Sodium Imp. F (EP), Ampicillin Imp. F (EP), (2RS,4S)-2-[[[(2R)-2-Amino-2-phenylacetyl]amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid, Penilloic Acids of Ampicillin, Ampicillin Sodium Impurity F, Ampicillin Impurity F
InChI Formula InChI=1S/C15H21N3O3S/c1-15(2)12(14(20)21)18-10(22-15)8-17-13(19)11(16)9-6-4-3-5-7-9/h3-7,10-12,18H,8,16H2,1-2H3,(H,17,19)(H,20,21)/t10?,11-,12+/m1/s1
SMILES CC1(C)SC(CNC(=O)[C@H](N)c2ccccc2)N[C@H]1C(=O)O
IUPAC Name (4S)-2-[[[(2R)-2-amino-2-phenylacetyl]amino]methyl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylic acid
Molecular Weight (g/mol) 323.41
Quantity 25 mg
Formula Weight 323.1304 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.