Learn More
(2S)-2-(3-Aminopropyl)-5-(2,5-difluorophenyl)-N-methoxy-N-methyl-2-phenyl-1,3,4-thiadiazole-3(2H)-carboxamide Hydrochloride, TRC
Product Code. 30314500
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314500 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30314500 Supplier Toronto Research Chemicals Supplier No. TRCA62832810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2S)-2-(3-Aminopropyl)-5-(2,5-difluorophenyl)-N-methoxy-N-methyl-2-phenyl-1,3,4-thiadiazole-3(2H)-carboxamide Hydrochloride
CAS 1385020-40-5
Molecular Formula C20H22F2N4O2S . (HCl)
Quantity 10 mg
Synonym ARRY 520 Hydrochloride, Filanesib Hydrochloride
InChI Formula InChI=1S/C20H22F2N4O2S.ClH/c1-25(28-2)19(27)26-20(11-6-12-23,14-7-4-3-5-8-14)29-18(24-26)16-13-15(21)9-10-17(16)22;/h3-5,7-10,13H,6,11-12,23H2,1-2H3;1H/t20-;/m0./s1
SMILES Cl.CON(C)C(=O)N1N=C(S[C@@]1(CCCN)c2ccccc2)c3cc(F)ccc3F
IUPAC Name (2S)-2-(3-aminopropyl)-5-(2,5-difluorophenyl)-N-methoxy-N-methyl-2-phenyl-1,3,4-thiadiazole-3-carboxamide;hydrochloride
Molecular Weight (g/mol) 456.9
Formula Weight 456.1198