Learn More
(2S)-2-(3-Aminopropyl)-5-(2,5-difluorophenyl)-N-methoxy-N-methyl-2-phenyl-1,3,4-thiadiazole-3(2H)-carboxamide Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (2S)-2-(3-Aminopropyl)-5-(2,5-difluorophenyl)-N-methoxy-N-methyl-2-phenyl-1,3,4-thiadiazole-3(2H)-carboxamide Hydrochloride |
| CAS | 1385020-40-5 |
| Molecular Formula | C20H22F2N4O2S . (HCl) |
| Quantity | 50 mg |
| Synonym | ARRY 520 Hydrochloride, Filanesib Hydrochloride |
| InChI Formula | InChI=1S/C20H22F2N4O2S.ClH/c1-25(28-2)19(27)26-20(11-6-12-23,14-7-4-3-5-8-14)29-18(24-26)16-13-15(21)9-10-17(16)22;/h3-5,7-10,13H,6,11-12,23H2,1-2H3;1H/t20-;/m0./s1 |
| SMILES | Cl.CON(C)C(=O)N1N=C(S[C@@]1(CCCN)c2ccccc2)c3cc(F)ccc3F |
| IUPAC Name | (2S)-2-(3-aminopropyl)-5-(2,5-difluorophenyl)-N-methoxy-N-methyl-2-phenyl-1,3,4-thiadiazole-3-carboxamide;hydrochloride |
| Molecular Weight (g/mol) | 456.9 |
| Formula Weight | 456.1198 |