missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S)-2-[(3S,8aR)-3-(4-Aminobutyl)-1,4-dioxohexahydropyrrolo[1,2-a]pyrazin-2(1H)-yl]-4-phenylbutanoic Acid (R,S,S-Diketopiperazine), Mikromol™
Description
(2S)-2-[(3S,8aR)-3-(4-Aminobutyl)-1,4-dioxohexahydropyrrolo[1,2-a]pyrazin-2(1H)-yl]-4-phenylbutanoic Acid (R,S,S-Diketopiperazine)
Specifications
Specifications
| Chemical Name or Material | Lisinopril Diketopiperazine |
| CAS | 219677-82-4 |
| Analyte or Component Names | Lisinopril Diketopiperazine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H29 N3 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Lisinopril Dihydrate Imp. D (EP), Lisinopril Imp. D (EP), Lisinopril diketopiperazine, (2S)-2-[(3S,8aR)-3-(4-Aminobutyl)-1,4-dioxohexahydropyrrolo[1,2-a]pyrazin-2(H)-yl]-4-phenylbutanoic acid, Lisinopril Dihydrate Impurity D, Lisinopril Impurity D |
| InChI Formula | InChI=1S/C21H29N3O4/c22-13-5-4-9-17-19(25)23-14-6-10-16(23)20(26)24(17)18(21(27)28)12-11-15-7-2-1-3-8-15/h1-3,7-8,16-18H,4-6,9-14,22H2,(H,27,28)/t16-,17+,18+/m1/s1 |
| SMILES | [C@H](CCC1=CC=CC=C1)(C(O)=O)N2[C@@H](CCCCN)C(=O)N3[C@@](C2=O)(CCC3)[H] |
| IUPAC Name | (2S)-2-[(3S,8aR)-3-(4-aminobutyl)-1,4-dioxo-6,7,8,8a-tetrahydro-3H-pyrrolo[1,2-a]pyrazin-2-yl]-4-phenylbutanoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?