missing translation for 'onlineSavingsMsg'
Learn More

(2S)-2-[[4-[[(2,4-Diaminopteridin-6-yl)methyl]amino]benzoyl]amino]pentanedioic Acid (4-Aminofolic Acid, Aminopterin), Mikromol™

Product Code. 30052721
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30052721

Brand: Mikromol MM0040.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2S)-2-[[4-[[(2,4-Diaminopteridin-6-yl)methyl]amino]benzoyl]amino]pentanedioic Acid (4-Aminofolic Acid, Aminopterin)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aminopterin
CAS 54-62-6
Analyte or Component Names Aminopterin
Shipping Condition Room Temperature
Molecular Formula C19 H20 N8 O5
Certifications/Compliance ISO 9001
Synonym L-Glutamic acid, N-[4-[[(2,4-diamino-6-pteridinyl)methyl]amino]benzoyl]-, Glutamic acid, N-[p-[[(2,4-diamino-6-pteridinyl)methyl]amino]benzoyl]-, L- (8CI), N-[4-[[(2,4-Diamino-6-pteridinyl)methyl]amino]benzoyl]-L-glutamic acid, 4-Aminofolic acid, 4-Aminopteroylglutamic acid, APGA, Aminopterin, Aminopterine, NSC 739, Pteramina, (S)-2-{4-[(2,4-Diaminopteridin-6-yl)methylamino]benzamido}pentanedioic acid, Methotrexate Imp. B (EP), (2S)-2-[[4-[[(2,4-Diaminopteridin-6-yl)methyl]amino]benzoyl]amino]pentanedioic Acid, Methotrexate Related Compound B, Methotrexate Impurity B
InChI Formula InChI=1S/C19H20N8O5/c20-15-14-16(27-19(21)26-15)23-8-11(24-14)7-22-10-3-1-9(2-4-10)17(30)25-12(18(31)32)5-6-13(28)29/h1-4,8,12,22H,5-7H2,(H,25,30)(H,28,29)(H,31,32)(H4,20,21,23,26,27)/t12-/m0/s1
SMILES Nc1nc(N)c2nc(CNc3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O)cnc2n1
IUPAC Name (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
Molecular Weight (g/mol) 440.41
Quantity 25 mg
Formula Weight 440.1557 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.