missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S)-2-[[4-[[(2,4-Diaminopteridin-6-yl)methyl]methylamino]benzoyl]amino]-5-methoxy-5-oxopentanoic Acid (Methotrexate 5-Methyl Ester), Mikromol™
Description
(2S)-2-[[4-[[(2,4-Diaminopteridin-6-yl)methyl]methylamino]benzoyl]amino]-5-methoxy-5-oxopentanoic Acid (Methotrexate 5-Methyl Ester)
Specifications
Specifications
| Chemical Name or Material | Methotrexate 5-Methyl Ester |
| CAS | 67022-39-3 |
| Analyte or Component Names | Methotrexate 5-Methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H24 N8 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | L-Glutamic acid, N-[4-[[(2,4-diamino-6-pteridinyl)methyl]methylamino]benzoyl]-, 5-methyl ester, Methotrexate 5-Methyl Ester, Methotrexate Imp. H (EP), Methotrexate Related Compound H, Methotrexate Impurity H |
| InChI Formula | InChI=1S/C21H24N8O5/c1-29(10-12-9-24-18-16(25-12)17(22)27-21(23)28-18)13-5-3-11(4-6-13)19(31)26-14(20(32)33)7-8-15(30)34-2/h3-6,9,14H,7-8,10H2,1-2H3,(H,26,31)(H,32,33)(H4,22,23,24,27,28)/t14-/m0/s1 |
| SMILES | COC(=O)CC[C@H](NC(=O)c1ccc(cc1)N(C)Cc2cnc3nc(N)nc(N)c3n2)C(=O)O |
| IUPAC Name | (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]-5-methoxy-5-oxopentanoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?