missing translation for 'onlineSavingsMsg'
Learn More

(2S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3-iodophenyl]propanoic Acid (3,3',5'-Triiodo-L-Thyronine), Mikromol™

Product Code. 18023950
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18023950 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18023950 Supplier Mikromol Supplier No. MM0694.100025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3-iodophenyl]propanoic Acid (3,3',5'-Triiodo-L-Thyronine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,3',5'-Triiodo-L-thyronine (Reverse T3)
CAS 5817-39-0
Analyte or Component Names 3,3',5'-Triiodo-L-thyronine (Reverse T3)
Shipping Condition Room Temperature
Molecular Formula C15 H12 I3 N O4
Certifications/Compliance ISO 9001
Synonym O-(4-Hydroxy-3,5-diiodophenyl)-3-iodo-L-tyrosine, 3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3-iodophenyl]-L-alanine, 3,3',5'-L-Triiodothyronine, 3,3',5'-T3, 3,3',5'-Triiodothyronine, Isoliothyronine, Reverse L-triiodothyronine, Reverse T3, Reverse triiodothyronine, rT3, 3,3',5-Triiodo-L-thyronine (T3), Levothyroxine Sodium Impurity K, Levothyroxine Impurity K
InChI Formula InChI=1S/C15H12I3NO4/c16-9-3-7(4-12(19)15(21)22)1-2-13(9)23-8-5-10(17)14(20)11(18)6-8/h1-3,5-6,12,20H,4,19H2,(H,21,22)/t12-/m0/s1
SMILES N[C@@H](Cc1ccc(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(=O)O
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3-iodophenyl]propanoic acid
Molecular Weight (g/mol) 650.97
Quantity 25 mg
Formula Weight 650.79 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.