Learn More
(2S)-2-[[(phenylmethoxy)carbonyl]amino]-4-Pentenoic acid, TRC
Specifications
Specifications
| Chemical Name or Material | (2S)-2-(((phenylmethoxy)carbonyl)amino)-4-Pentenoic acid |
| CAS | 78553-51-2 |
| Molecular Formula | C13H15NO4 |
| Quantity | 100 mg |
| Synonym | 4-Pentenoic acid','2-[[(phenylmethoxy)carbonyl]amino]-','(S)-, (2S)-2-[[(Phenylmethoxy)carbonyl]amino]-4-pentenoic acid, (S)-2-[[(Benzyloxy)carbonyl]amino]pent-4-enoic acid |
| InChI Formula | InChI=1S/C13H15NO4/c1-2-6-11(12(15)16)14-13(17)18-9-10-7-4-3-5-8-10/h2-5,7-8,11H,1,6,9H2,(H,14,17)(H,15,16)/t11-/m0/s1 |
| SMILES | OC(=O)[C@H](CC=C)NC(=O)OCc1ccccc1 |
| IUPAC Name | (2S)-2-(phenylmethoxycarbonylamino)pent-4-enoic acid |
| Molecular Weight (g/mol) | 249.26 |
| Formula Weight | 249.1001 |