missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S,3aS,6aS)-Octahydrocyclopenta[b]pyrrole-2-carboxylic Acid Phenylmethyl Ester Hydrochloride, Mikromol™
Description
(2S,3aS,6aS)-Octahydrocyclopenta[b]pyrrole-2-carboxylic Acid Phenylmethyl Ester Hydrochloride, Mikromol
Specifications
Specifications
| Chemical Name or Material | (2S,3aS,6aS)-Octahydrocyclopenta[b]pyrrole-2-carboxylic Acid Phenylmethyl Ester Hydrochloride |
| CAS | 87269-87-2 |
| Analyte or Component Names | (2S,3aS,6aS)-Octahydrocyclopenta[b]pyrrole-2-carboxylic Acid Phenylmethyl Ester Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H19 N O2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | (2S,3aS,6aS)-Octahydrocyclopenta[b]pyrrole-2-carboxylic Acid Phenylmethyl Ester Hydrochloride |
| InChI Formula | InChI=1S/C15H19NO2.ClH/c17-15(18-10-11-5-2-1-3-6-11)14-9-12-7-4-8-13(12)16-14;/h1-3,5-6,12-14,16H,4,7-10H2;1H/t12-,13-,14-;/m0./s1 |
| Solution Type | Neat |
| SMILES | Cl.O=C(OCc1ccccc1)[C@@H]2C[C@@H]3CCC[C@@H]3N2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?