missing translation for 'onlineSavingsMsg'
Learn More

(2S,3aS,7aS)-1-Fmoc-octahydroindole-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number 15448336
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
130309-37-4
C24H25NO4
391.467
MFCD00191201
JBZXLQHJZHITMW-RXYZOABWSA-N
fmoc-oic-oh, 2s,3as,7as-1-9h-fluoren-9-yl methoxy carbonyl octahydro-1h-indole-2-carboxylic acid, fmoc-2s,3as,7as-octahydro-1h-indole-2-carboxylic acid, 2s,3as,7as-1-9h-fluoren-9-ylmethoxy carbonyl-octahydroindole-2-carboxylic acid, fmoc-l-octahydroindole-2-carboxylic acid, ambotzfaa1729, 2s pound not3as pound not7as-1-9h-fluoren-9-yl methoxy carbonyl, 2s,3as,7as-1-9h-fluoren-9-yl methoxy carbonyl-octahydro-1h-indole-2-carboxylic acid, 2s,3as,7as-1-9h-fluoren-9-ylmethoxycarbonyl-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid
2756165
(2S,3aS,7aS)-1-(9H-fluoren-9-ylmethoxycarbonyl)-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid
C1CCC2C(C1)CC(N2C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)C(=O)O
This item is not returnable. View return policy
130309-37-4
C24H25NO4
391.467
MFCD00191201
JBZXLQHJZHITMW-RXYZOABWSA-N
fmoc-oic-oh, 2s,3as,7as-1-9h-fluoren-9-yl methoxy carbonyl octahydro-1h-indole-2-carboxylic acid, fmoc-2s,3as,7as-octahydro-1h-indole-2-carboxylic acid, 2s,3as,7as-1-9h-fluoren-9-ylmethoxy carbonyl-octahydroindole-2-carboxylic acid, fmoc-l-octahydroindole-2-carboxylic acid, ambotzfaa1729, 2s pound not3as pound not7as-1-9h-fluoren-9-yl methoxy carbonyl, 2s,3as,7as-1-9h-fluoren-9-yl methoxy carbonyl-octahydro-1h-indole-2-carboxylic acid, 2s,3as,7as-1-9h-fluoren-9-ylmethoxycarbonyl-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid
2756165
(2S,3aS,7aS)-1-(9H-fluoren-9-ylmethoxycarbonyl)-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid
C1CCC2C(C1)CC(N2C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 1 g
Formula Weight 391.46
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (2S,3aS,7aS)-1-Fmoc-octahydroindole-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'