Learn More
(2S,3S)-2-[[(2Z)-2-(2-Amino-4-thiazolyl)-2-[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-3-(sulfoamino)butanoic Acid, TRC
Specifications
Specifications
| Analyte or Component Names | Open Ring Aztreonam |
| Shipping Condition | Room Temperature |
| InChI Formula | InChI=1S/C13H19N5O9S2/c1-5(18-29(24,25)26)7(10(20)21)16-9(19)8(6-4-28-12(14)15-6)17-27-13(2,3)11(22)23/h4-5,7,18H,1-3H3,(H2,14,15)(H,16,19)(H,20,21)(H,22,23)(H,24,25,26)/b17-8-/t5-,7-/m0/s1 |
| Solution Type | Neat |
| Industry Type | Forensics |
| Quantity | 25 mg |
| Formula Weight | 453.0624 |
| Chemical Name or Material | Open Ring Aztreonam |