Learn More
(2S,3S)-2-[[(2Z)-2-(2-Amino-4-thiazolyl)-2-[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-3-(sulfoamino)butanoic Acid, TRC
Product Code. 17963695
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963695 10 mg 10mg
17954045 25 mg 25mg
17952135 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17963695 Supplier Toronto Research Chemicals Supplier No. TRCA63041010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 87500-74-1
Molecular Formula C13 H19 N5 O9 S2
Molecular Weight (g/mol) 453.45
Synonym Butanoic acid, 2-[[(2Z)-2-(2-amino-4-thiazolyl)-2-[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-3-(sulfoamino)-, (2S,3S)-, Butanoic acid, 2-[[(2-amino-4-thiazolyl)[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-3-(sulfoamino)-, [S-[R*,R*-(Z)]]-, Butanoic acid, 2-[[(2Z)-(2-amino-4-thiazolyl)[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-3-(sulfoamino)-, (2S,3S)- (9CI), SQ 26992, Open Ring Aztreonam (USP)
IUPAC Name (2S,3S)-2-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(2-carboxypropan-2-yloxyimino)acetyl]amino]-3-(sulfoamino)butanoic acid
SMILES C[C@H](NS(=O)(=O)O)[C@H](NC(=O)\C(=N/OC(C)(C)C(=O)O)\c1csc(N)n1)C(=O)O
Specifications
Analyte or Component Names Open Ring Aztreonam
Shipping Condition Room Temperature
InChI Formula InChI=1S/C13H19N5O9S2/c1-5(18-29(24,25)26)7(10(20)21)16-9(19)8(6-4-28-12(14)15-6)17-27-13(2,3)11(22)23/h4-5,7,18H,1-3H3,(H2,14,15)(H,16,19)(H,20,21)(H,22,23)(H,24,25,26)/b17-8-/t5-,7-/m0/s1
Solution Type Neat
Industry Type Forensics
Quantity 10 mg
Formula Weight 453.0624
Chemical Name or Material Open Ring Aztreonam