Learn More
(2S)-4-Amino-2-hydroxybutanoic Acid, Mikromol™
Description
(2S)-4-Amino-2-hydroxybutanoic Acid
Specifications
Specifications
| Chemical Name or Material | (2S)-4-Amino-2-hydroxybutanoic Acid |
| CAS | 40371-51-5 |
| Analyte or Component Names | (2S)-4-Amino-2-hydroxybutanoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C4 H9 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Amikacin Sulfate Imp. I (EP), Amikacin Imp. I (EP), (2S)-4-Amino-2-hydroxybutanoic Acid, Butanoic acid, 4-amino-2-hydroxy-, (S)-, (2S)-4-Amino-2-hydroxybutanoic acid, (S)-4-Amino-2-hydroxybutanoic acid, (S)-4-Amino-2-hydroxybutyric acid, 4-Amino-(2S)-hydroxybutyric acid, L-(-)-4-Amino-2-hydroxybutyric acid, L-(-)-gamma-Amino-alpha-hydroxybutyric acid, L-4-Amino-2-hydroxybutyric acid, L-gamma-Amino-alpha-hydroxybutyric acid, Amikacin Impurity I, Amikacin Sulfate Impurity I, Amikacin Impurity I |
| InChI Formula | InChI=1S/C4H9NO3/c5-2-1-3(6)4(7)8/h3,6H,1-2,5H2,(H,7,8)/t3-/m0/s1 |
| SMILES | NCC[C@H](O)C(=O)O |
| IUPAC Name | (2S)-4-amino-2-hydroxybutanoic acid |
| Show More |