Learn More
(2S)-4-Amino-2-hydroxybutanoic Acid, Mikromol™
Product Code. 30052297
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052297 25 mg 25mg
30052286 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30052297 Supplier Mikromol Supplier No. MM1279.010025
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2S)-4-Amino-2-hydroxybutanoic Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2S)-4-Amino-2-hydroxybutanoic Acid
CAS 40371-51-5
Analyte or Component Names (2S)-4-Amino-2-hydroxybutanoic Acid
Shipping Condition Room Temperature
Molecular Formula C4 H9 N O3
Certifications/Compliance ISO 17025
Synonym Amikacin Sulfate Imp. I (EP), Amikacin Imp. I (EP), (2S)-4-Amino-2-hydroxybutanoic Acid, Butanoic acid, 4-amino-2-hydroxy-, (S)-, (2S)-4-Amino-2-hydroxybutanoic acid, (S)-4-Amino-2-hydroxybutanoic acid, (S)-4-Amino-2-hydroxybutyric acid, 4-Amino-(2S)-hydroxybutyric acid, L-(-)-4-Amino-2-hydroxybutyric acid, L-(-)-gamma-Amino-alpha-hydroxybutyric acid, L-4-Amino-2-hydroxybutyric acid, L-gamma-Amino-alpha-hydroxybutyric acid, Amikacin Impurity I, Amikacin Sulfate Impurity I, Amikacin Impurity I
InChI Formula InChI=1S/C4H9NO3/c5-2-1-3(6)4(7)8/h3,6H,1-2,5H2,(H,7,8)/t3-/m0/s1
SMILES NCC[C@H](O)C(=O)O
IUPAC Name (2S)-4-amino-2-hydroxybutanoic acid
Molecular Weight (g/mol) 119.12
Quantity 25 mg
Formula Weight 119.0582 g/mol