Learn More
2S,4R-Sacubitril, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
2S,4R-Sacubitril is the impurity of Sacubitril. Sacubitril is approved by the Food and Drug Administration for use in combination with valsartan for the treatment of patients with heart failure.
Specifications
Specifications
| Chemical Name or Material | 2S,4R-Sacubitril |
| CAS | 2307668-79-5 |
| Color | Colorless |
| Purity Grade Notes | Research |
| Molecular Formula | C25H31NO5 |
| Quantity | 5 mg |
| SMILES | [H]N(C(CC(C)C(O)=O)=O)[C@H](CC(C=C1)=CC=C1C2=CC=CC=C2)C[C@@H](C)C(OCC)=O |
| Molecular Weight (g/mol) | 425.52 |
| Formula Weight | 425.52 |
| Percent Purity | 99.12% |
| Show More |
Research purposes only