missing translation for 'onlineSavingsMsg'
Learn More

(2S,5R)-3,3-dimethyl-7-oxo-4-oxide-4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Diphenylmethyl Ester, TRC

Product Code. 30381407
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381407 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381407 Supplier Toronto Research Chemicals Supplier No. TRCD47423550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S,5R)-3,3-dimethyl-7-oxo-4-oxide-4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Diphenylmethyl Ester
CAS 87579-78-0
Molecular Formula C21H21NO4S
Quantity 50 mg
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-, diphenylmethyl ester, 4-oxide, (2S,5R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-, diphenylmethyl ester, 4-oxide, [2S-(2α,5α)]-, (2S,5R)-3,3-Dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid diphenylmethyl ester 4-oxide
InChI Formula InChI=1S/C21H21NO4S/c1-21(2)19(22-16(23)13-17(22)27(21)25)20(24)26-18(14-9-5-3-6-10-14)15-11-7-4-8-12-15/h3-12,17-19H,13H2,1-2H3/t17-,19+,27?/m1/s1
SMILES CC1(C)[C@@H](N2[C@@H](CC2=O)S1=O)C(=O)OC(c3ccccc3)c4ccccc4
IUPAC Name benzhydryl (2S,5R)-3,3-dimethyl-4,7-dioxo-4λ^{4}-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 383.46
Formula Weight 383.1191
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.