missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2Z)-(2-Aminothiazol-4-yl)(methoxyimino)acetic Acid, Mikromol™
Description
(2Z)-(2-Aminothiazol-4-yl)(methoxyimino)acetic Acid
Specifications
Specifications
| Chemical Name or Material | (2Z)-(2-Aminothiazol-4-yl)(methoxyimino)acetic Acid |
| CAS | 65872-41-5 |
| Analyte or Component Names | (2Z)-(2-Aminothiazol-4-yl)(methoxyimino)acetic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H7 N3 O3 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | (Z)-(2-Aminothiazol-4-yl)methoxyiminoacetic acid, (Z)-2-(2-Aminothiazol-4-yl)-2-methoxyiminoacetic acid, 2-(2-Amino-4-thiazolyl)-(Z)-2-methoxyiminoacetic acid, 2-(2-Aminothiazol-4-yl)-2-(Z)-methoxyiminoacetic acid, 2-(2-Aminothiazole-4-yl)-2-(Z)-methoxyiminoacetic acid, Thiazoximic acid, syn-2-(2-Aminothiazol-4-yl)-2-methoxyiminoacetic acid, syn-Thiazoximic acid, (2Z)-(2-Aminothiazol-4-yl)(methoxyimino)acetic acid, Cefepime Dihydrochloride Monohydrate Imp. D (EP), Cefepime Imp. D (EP), Cefepime USP Related Compound D, Cefepime USP RC D, 4-Thiazoleacetic acid, 2-amino-alpha-(methoxyimino)-, (alphaZ)-, 4-Thiazoleacetic acid, 2-amino-alpha-(methoxyimino)-, (Z)-, (Z)-(2-Aminothiazol-4-yl)-alpha-(methoxyimino)acetic acid, (Z)-2-Amino-alpha-(methoxyimino)-4-thiazoleacetic acid, Cefepime Related Compound D, Cefepime Dihydrochloride Monohydrate Impurity D, Cefepime Impurity D |
| InChI Formula | InChI=1S/C6H7N3O3S/c1-12-9-4(5(10)11)3-2-13-6(7)8-3/h2H,1H3,(H2,7,8)(H,10,11)/b9-4- |
| SMILES | CO\N=C(/C(=O)O)\c1csc(N)n1 |
| IUPAC Name | (2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?