Learn More
Alfa Aesar™ 3-(1-Piperidinylmethyl)indole, 95%
Brand:  Alfa Aesar™ H63256.03
Additional Details : CAS : 5355-42-0 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15439858
  • € 207.00 / 1g
Estimated Shipment date
from Supplier 30-06-2020
Add to basket
Description
Description
Applications
3-(1-Piperidinylmethyl)indole, is used as a pharmaceutical intermediate.
Solubility
Freely soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Chemical Identifiers
5355-42-0
214.312
HPKXPFRRVVWAPD-UHFFFAOYSA-N
21454
C1CCN(CC1)CC2=CNC3=CC=CC=C32
C14H18N2
MFCD00458412
n-skatylpiperidine, 3-piperidin-1-yl methyl-1h-indole, 3-piperidinomethyl indole, indole, 3-piperidinomethyl, 3-piperidin-1-ylmethyl-1h-indole, chembl57535, 3-piperidinomethyl-1h-indole, 3-1-piperidinylmethyl-1h-indole, 3-piperidylmethyl indole
3-(piperidin-1-ylmethyl)-1H-indole
Specifications
3-(1-Piperidinylmethyl)indole
1g
C14H18N2
176547
Freely soluble in water.
C1CCN(CC1)CC2=CNC3=CC=CC=C32
214.312
214.3
95%
Tan to White
5355-42-0
MFCD00458412
n-skatylpiperidine, 3-piperidin-1-yl methyl-1h-indole, 3-piperidinomethyl indole, indole, 3-piperidinomethyl, 3-piperidin-1-ylmethyl-1h-indole, chembl57535, 3-piperidinomethyl-1h-indole, 3-1-piperidinylmethyl-1h-indole, 3-piperidylmethyl indole
HPKXPFRRVVWAPD-UHFFFAOYSA-N
3-(piperidin-1-ylmethyl)-1H-indole
21454
Solid