missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methyl-1H-imidazol-1-yl)aniline, 97%, Thermo Scientific™

Catalog Number p-3541088
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
184098-19-9
C10H11N3
173.219
MFCD09702403
ZDHNDCMKVXWWLA-UHFFFAOYSA-N
3-2-methyl-1h-imidazol-1-yl aniline, 3-2-methylimidazol-1-yl aniline, 3-2-methyl-1h-imidazol-1-yl-benzenamine, 3-2-methylimidazolyl phenylamine, benzenamine,3-2-methyl-1h-imidazol-1-yl
12797139
3-(2-methylimidazol-1-yl)aniline
CC1=NC=CN1C2=CC(=CC=C2)N
This item is not returnable. View return policy
184098-19-9
C10H11N3
173.219
MFCD09702403
ZDHNDCMKVXWWLA-UHFFFAOYSA-N
3-2-methyl-1h-imidazol-1-yl aniline, 3-2-methylimidazol-1-yl aniline, 3-2-methyl-1h-imidazol-1-yl-benzenamine, 3-2-methylimidazolyl phenylamine, benzenamine,3-2-methyl-1h-imidazol-1-yl
12797139
3-(2-methylimidazol-1-yl)aniline
CC1=NC=CN1C2=CC(=CC=C2)N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 3-(2-Methyl-1H-imidazol-1-yl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'