missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Naphthyl)-D-alanine, 95%, Thermo Scientific Chemicals

Catalog Number 15440269
61.13 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
76985-09-6
C13H13NO2
215.25
MFCD00038546
JPZXHKDZASGCLU-GFCCVEGCSA-N
3-2-naphthyl-d-alanine, h-d-2-nal-oh, r-2-amino-3-naphthalen-2-yl propanoic acid, r-2-amino-3-naphthalene-2-yl-propionic acid, d-2-naphthylalanine, d-3-2-naphthyl-alanine, unii-571v312ymy, 2-d-naphthylalanine, 2r-2-amino-3-2-naphthyl propanoic acid, 2r-2-amino-3-naphthalen-2-ylpropanoic acid
2733661
(2R)-2-amino-3-naphthalen-2-ylpropanoic acid
[NH3+][C@H](CC1=CC=C2C=CC=CC2=C1)C([O-])=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
76985-09-6
C13H13NO2
215.25
MFCD00038546
JPZXHKDZASGCLU-GFCCVEGCSA-N
3-2-naphthyl-d-alanine, h-d-2-nal-oh, r-2-amino-3-naphthalen-2-yl propanoic acid, r-2-amino-3-naphthalene-2-yl-propionic acid, d-2-naphthylalanine, d-3-2-naphthyl-alanine, unii-571v312ymy, 2-d-naphthylalanine, 2r-2-amino-3-2-naphthyl propanoic acid, 2r-2-amino-3-naphthalen-2-ylpropanoic acid
2733661
(2R)-2-amino-3-naphthalen-2-ylpropanoic acid
[NH3+][C@H](CC1=CC=C2C=CC=CC2=C1)C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Beilstein 4352384
Formula Weight 215.25
Percent Purity 95%
Quantity 1 g
Physical Form Solid
Chemical Name or Material 3-(2-Naphthyl)-D-alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'