Learn More
(3,3,4,4,4-d5)-(E/Z)-1-[4-(2-Chloroethoxy)phenyl]-1-[4-(t-butyldimethylsilyloxy)phenyl]-2-phenyl-1-butene, TRC
Specifications
Specifications
| Chemical Name or Material | (3,3,4,4,4-d5)-(E/Z)-1-[4-(2-Chloroethoxy)phenyl]-1-[4-(t-butyldimethylsilyloxy)phenyl]-2-phenyl-1-butene |
| CAS | 1020719-26-9 |
| Molecular Formula | C30H32D5ClO2Si |
| Quantity | 250 mg |
| Synonym | 1-(2-Chloroethoxy)-4-[1-[4-[[(1,1-dimethylethyl)dimethylsilyl]oxy]phenyl]-2-phenyl-1-buten-1-yl-3,3,4,4,4-d5]benzene |
| InChI Formula | InChI=1S/C30H37ClO2Si/c1-7-28(23-11-9-8-10-12-23)29(24-13-17-26(18-14-24)32-22-21-31)25-15-19-27(20-16-25)33-34(5,6)30(2,3)4/h8-20H,7,21-22H2,1-6H3/b29-28+/i1D3,7D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])\C(=C(\c1ccc(OCCCl)cc1)/c2ccc(O[Si](C)(C)C(C)(C)C)cc2)\c3ccccc3 |
| IUPAC Name | tert-butyl-[4-[(E)-1-[4-(2-chloroethoxy)phenyl]-3,3,4,4,4-pentadeuterio-2-phenylbut-1-enyl]phenoxy]-dimethylsilane |
| Molecular Weight (g/mol) | 498.18 |
| Formula Weight | 497.2565 |