Learn More
3,3’,5’-Triiodo-L-thyronine, TRC
Product Code. 18033220
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18033220 10 mg 10mg
18053220 100 mg 100mg
18001230 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18033220 Supplier Toronto Research Chemicals Supplier No. TRCT79535010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5817-39-0
Molecular Formula C15 H12 I3 N O4
Molecular Weight (g/mol) 650.97
Synonym O-(4-Hydroxy-3,5-diiodophenyl)-3-iodo-L-tyrosine, 3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3-iodophenyl]-L-alanine, 3,3',5'-L-Triiodothyronine, 3,3',5'-T3, 3,3',5'-Triiodothyronine, Isoliothyronine, Reverse L-triiodothyronine, Reverse T3, Reverse triiodothyronine, rT3
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3-iodophenyl]propanoic acid
SMILES N[C@@H](Cc1ccc(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(=O)O
Specifications
InChI Formula InChI=1S/C15H12I3NO4/c16-9-3-7(4-12(19)15(21)22)1-2-13(9)23-8-5-10(17)14(20)11(18)6-8/h1-3,5-6,12,20H,4,19H2,(H,21,22)/t12-/m0/s1
Quantity 10 mg
Formula Weight 650.79
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material 3,3',5'-Triiodo-L-thyronine (Reverse T3)
Grade HPLC