missing translation for 'onlineSavingsMsg'
Learn More

3,3’,5’-Triiodo-L-thyronine, TRC

Product Code. 18053220
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18053220 100 mg 100mg
18033220 10 mg 10mg
18001230 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18053220 Supplier Toronto Research Chemicals Supplier No. TRCT795350100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5817-39-0
Molecular Formula C15 H12 I3 N O4
Molecular Weight (g/mol) 650.97
Synonym O-(4-Hydroxy-3,5-diiodophenyl)-3-iodo-L-tyrosine, 3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3-iodophenyl]-L-alanine, 3,3',5'-L-Triiodothyronine, 3,3',5'-T3, 3,3',5'-Triiodothyronine, Isoliothyronine, Reverse L-triiodothyronine, Reverse T3, Reverse triiodothyronine, rT3
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3-iodophenyl]propanoic acid
SMILES N[C@@H](Cc1ccc(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(=O)O

Specifications

InChI Formula InChI=1S/C15H12I3NO4/c16-9-3-7(4-12(19)15(21)22)1-2-13(9)23-8-5-10(17)14(20)11(18)6-8/h1-3,5-6,12,20H,4,19H2,(H,21,22)/t12-/m0/s1
Quantity 100 mg
Formula Weight 650.79
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material 3,3',5'-Triiodo-L-thyronine (Reverse T3)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.