missing translation for 'onlineSavingsMsg'
Learn More

3-[3-Acetyl-4-[[(RS)-oxiranyl]-methoxy]phenyl]-1,1-diethylurea, Mikromol™

Product Code. 30049600
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30049600

Brand: Mikromol MM0128.06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-[3-Acetyl-4-[[(RS)-oxiranyl]-methoxy]phenyl]-1,1-diethylurea
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[3-Acetyl-4-[((RS)-oxiranyl)-methoxy]phenyl]-1,1-diethylurea
CAS 56980-98-4
Analyte or Component Names 3-[3-Acetyl-4-[((RS)-oxiranyl)-methoxy]phenyl]-1,1-diethylurea
Shipping Condition Ice pack (-20°C)
Molecular Formula C16 H22 N2 O4
Certifications/Compliance ISO 9001
Synonym Celiprolol Hydrochloride Imp. G (EP), Celiprolol Imp. G (EP), 3-[3-Acetyl-4-[[(RS)-oxiranyl]-methoxy]phenyl]-1,1-diethylurea, Celiprolol Hydrochloride Impurity G, Celiprolol Impurity G
InChI Formula InChI=1S/C16H22N2O4/c1-4-18(5-2)16(20)17-12-6-7-15(14(8-12)11(3)19)22-10-13-9-21-13/h6-8,13H,4-5,9-10H2,1-3H3,(H,17,20)
SMILES CCN(CC)C(=O)Nc1ccc(OCC2CO2)c(c1)C(=O)C
IUPAC Name 3-[3-acetyl-4-(oxiran-2-ylmethoxy)phenyl]-1,1-diethylurea
Molecular Weight (g/mol) 306.36
Quantity 100 mg
Formula Weight 306.158 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.