Learn More
3,3-Azo-1-butanol, TRC
Specifications
Specifications
| Chemical Name or Material | 3,3-Azo-1-butanol |
| CAS | 25055-82-7 |
| Molecular Formula | C4 H8 N2 O |
| Quantity | 100 mg |
| Synonym | 3-Methyl-3-azibutanol, 3H-Diazirine-3-ethanol |
| InChI Formula | InChI=1S/C4H8N2O/c1-4(2-3-7)5-6-4/h7H,2-3H2,1H3 |
| SMILES | CC1(CCO)N=N1 |
| IUPAC Name | 2-(3-methyldiazirin-3-yl)ethanol |
| Molecular Weight (g/mol) | 100.12 |
| Formula Weight | 100.0637 |