Learn More
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass Filter Crucibles
€ 21.31 - € 36.94
Specifications
Description | 3.3 Borosilicate Glass |
---|---|
Material | Borosilicate Glass |
Product Type Specs | Filter crucible |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11556764
|
Lenz Laborglasinstrumente™
07185323 |
15 mL |
€ 21.31
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
10621902
|
Lenz Laborglasinstrumente™
07185430 |
30 mL |
€ 24.96
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
10796761
|
Lenz Laborglasinstrumente™
07185440 |
50 mL |
€ 36.94
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
Specifications
3.3 Borosilicate Glass | |
Borosilicate Glass | |
Filter crucible |