Learn More
3,3'-[Carbonylbis[imino(2-methyl-4,1-phenylene)-2,1-diazenediyl]]bis-1,5-naphthalenedisulfonic Acid Sodium Salt (Technical Grade), TRC
Product Code. 30476481
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476481 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30476481 Supplier Toronto Research Chemicals Supplier No. TRCN3784432.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3214-47-9
Molecular Formula C35H24N6Na4O13S4
Molecular Weight (g/mol) 956.82
Synonym C.I. Direct Yellow 50 (7CI), C.I. Direct Yellow 50 Tetrasodium Salt (8CI), Abcol Direct Yellow RLSW, C.I. 29025, Direct Fast Yellow LR, Direct Fast Yellow R, Direct Lightfast Yellow K, Direct Lightfast Yellow RS, Direct Yellow 50, Helion Yellow RL, Isma Fast Yellow RL, Levacell Fast Yellow R, Orbantin Yellow R, Pergasol Yellow 2R, Pyrazol Fast Yellow RSW, Sirius Light Yellow R, Sirius Light Yellow R Extra, Sirius Supra Yellow R, Sirius Yellow R, Solar Flavine RN, Solophenyl Yellow AGFL, Tubantin Yellow 3R
IUPAC Name tetrasodium;3-[(E)-[4-[[4-[(E)-(4,8-disulfonatonaphthalen-2-yl)diazenyl]-3-methylphenyl]carbamoylamino]-2-methylphenyl]diazenyl]naphthalene-1,5-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].Cc1cc(NC(=O)Nc2ccc(N=Nc3cc(c4cccc(c4c3)S(=O)(=O)[O-])S(=O)(=O)[O-])c(C)c2)ccc1N=Nc5cc(c6cccc(c6c5)S(=O)(=O)[O-])S(=O)(=O)[O-]
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C35H28N6O13S4.4Na/c1-19-13-21(9-11-29(19)40-38-23-15-27-25(33(17-23)57(49,50)51)5-3-7-31(27)55(43,44)45)36-35(42)37-22-10-12-30(20(2)14-22)41-39-24-16-28-26(34(18-24)58(52,53)54)6-4-8-32(28)56(46,47)48;;;;/h3-18H,1-2H3,(H2,36,37,42)(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4
Formula Weight 955.9875
Chemical Name or Material 3,3'-[Carbonylbis(imino(2-methyl-4,1-phenylene)-2,1-diazenediyl)]bis-1,5-naphthalenedisulfonic Acid Sodium Salt (Technical Grade)