Learn More
3,3'-[Carbonylbis[imino(2-methyl-4,1-phenylene)-2,1-diazenediyl]]bis-1,5-naphthalenedisulfonic Acid Sodium Salt (Technical Grade), TRC
Product Code. 30180228
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180228 5 g 5g
30180079 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30180228 Supplier Toronto Research Chemicals Supplier No. TRCN3784435G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3214-47-9
Molecular Formula C35H24N6Na4O13S4
Molecular Weight (g/mol) 956.82
Synonym C.I. Direct Yellow 50 (7CI);C.I. Direct Yellow 50 Tetrasodium Salt (8CI);Abcol Direct Yellow RLSW;C.I. 29025;Direct Fast Yellow LR;Direct Fast Yellow R;Direct Lightfast Yellow K;Direct Lightfast Yellow RS;Direct Yellow 50;Helion Yellow RL;Isma Fast Yellow RL;Levacell Fast Yellow R;Orbantin Yellow R;Pergasol Yellow 2R;Pyrazol Fast Yellow RSW;Sirius Light Yellow R;Sirius Light Yellow R Extra;Sirius Supra Yellow R;Sirius Yellow R;Solar Flavine RN;Solophenyl Yellow AGFL;Tubantin Yellow 3R;
IUPAC Name tetrasodium;3-[(E)-[4-[[4-[(E)-(4,8-disulfonatonaphthalen-2-yl)diazenyl]-3-methylphenyl]carbamoylamino]-2-methylphenyl]diazenyl]naphthalene-1,5-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].Cc1cc(NC(=O)Nc2ccc(N=Nc3cc(c4cccc(c4c3)S(=O)(=O)[O-])S(=O)(=O)[O-])c(C)c2)ccc1N=Nc5cc(c6cccc(c6c5)S(=O)(=O)[O-])S(=O)(=O)[O-]
Specifications
Quantity 5 g
InChI Formula InChI=1S/C35H28N6O13S4.4Na/c1-19-13-21(9-11-29(19)40-38-23-15-27-25(33(17-23)57(49,50)51)5-3-7-31(27)55(43,44)45)36-35(42)37-22-10-12-30(20(2)14-22)41-39-24-16-28-26(34(18-24)58(52,53)54)6-4-8-32(28)56(46,47)48;;;;/h3-18H,1-2H3,(H2,36,37,42)(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4
Formula Weight 955.9875
Chemical Name or Material 3,3'-[Carbonylbis(imino(2-methyl-4,1-phenylene)-2,1-diazenediyl)]bis-1,5-naphthalenedisulfonic Acid Sodium Salt (Technical Grade)