missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(3-Chlorophenyl)-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63510.06

Additional Details : CAS : 42351-67-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15440249

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

42351-67-7
226.678
OQIMHAMJYXODDN-UHFFFAOYSA-N
2560555
C1C(=O)N(C(=S)N1)C2=CC(=CC=C2)Cl
C9H7ClN2OS
MFCD04048637
1-3-chlorophenyl-2-sulfanyl-4,5-dihydro-1h-imidazol-5-one, 3-3-chlorophenyl-2-mercapto-3,5-dihydro-4h-imidazol-4-one, 3-3-chlorophenyl-2-sulfanylideneimidazolidin-4-one, 3-3-chlorophenyl-2-thioxoimidazolidin-4-one, 3-3-chlorophenyl-2-thioxo-1,3-diazolidin-4-one
3-(3-chlorophenyl)-2-sulfanylideneimidazolidin-4-one

Specifications

3-(3-Chlorophenyl)-2-thiohydantoin
42351-67-7
MFCD04048637
1-3-chlorophenyl-2-sulfanyl-4,5-dihydro-1h-imidazol-5-one, 3-3-chlorophenyl-2-mercapto-3,5-dihydro-4h-imidazol-4-one, 3-3-chlorophenyl-2-sulfanylideneimidazolidin-4-one, 3-3-chlorophenyl-2-thioxoimidazolidin-4-one, 3-3-chlorophenyl-2-thioxo-1,3-diazolidin-4-one
C1C(=O)N(C(=S)N1)C2=CC(=CC=C2)Cl
226.678
226.68
95%
5g
C9H7ClN2OS
786006
OQIMHAMJYXODDN-UHFFFAOYSA-N
3-(3-chlorophenyl)-2-sulfanylideneimidazolidin-4-one
2560555
Solid
Videos
Documents