Learn More
3,3-Diethoxypropionitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 3,3-Diethoxypropionitrile |
| CAS | 2032-34-0 |
| Molecular Formula | C7H13NO2 |
| Quantity | 10 g |
| Synonym | 3,3-Diethoxypropanenitrile, Cyanoacetaldehyde Diethyl Acetal, Malonaldehydenitrile Diethyl Acetal |
| InChI Formula | InChI=1S/C7H13NO2/c1-3-9-7(5-6-8)10-4-2/h7H,3-5H2,1-2H3 |
| SMILES | CCOC(CC#N)OCC |
| Molecular Weight (g/mol) | 143.18 |
| Formula Weight | 143.0946 |