Learn More
3,3-Difluorocyclobutanecarboxylic acid, 97%, Thermo Scientific™
Catalog Number p-7077382
Click to view available options
Quantity:
250 mg
1 g
107496-54-8
C5H6F2O2
136.1
PLRCVBKYFLWAAT-UHFFFAOYSA-N
3,3-difluorocyclobutanecarboxylic acid, 3,3-difluoro-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3,3-difluoro, cyclobutanecarboxylicacid, 3,3-difluoro, 3,3-difluorocyclobutanecarboxylicacid, acmc-209yhn, 3,3-difluorocyclopropyl methanol, 3,3-difluorocyclobutane carboxylic acid, 3,3-difluorocyclobutane-carboxylic acid, 3,3-difluoro-cyclobutane-1-carboxylic acid
13777789
3,3-difluorocyclobutane-1-carboxylic acid
C1C(CC1(F)F)C(=O)O
107496-54-8
C5H6F2O2
136.1
PLRCVBKYFLWAAT-UHFFFAOYSA-N
3,3-difluorocyclobutanecarboxylic acid, 3,3-difluoro-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3,3-difluoro, cyclobutanecarboxylicacid, 3,3-difluoro, 3,3-difluorocyclobutanecarboxylicacid, acmc-209yhn, 3,3-difluorocyclopropyl methanol, 3,3-difluorocyclobutane carboxylic acid, 3,3-difluorocyclobutane-carboxylic acid, 3,3-difluoro-cyclobutane-1-carboxylic acid
13777789
3,3-difluorocyclobutane-1-carboxylic acid
C1C(CC1(F)F)C(=O)O
Melting Point 50.0°C to 52.5°C
Density 1.3700g/mL
Boiling Point 205.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Quantity 250 mg
Specific Gravity 1.37
Formula Weight 136.1
Percent Purity 97%
Chemical Name or Material 3,3-Difluorocyclobutanecarboxylic acid
compliance-icons
Product Identifier
  • 3,3-Difluorocyclobutanecarboxylic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.