missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dimethyldiphenylamine, 98%, Thermo Scientific Chemicals

Catalog Number p-7025999
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
626-13-1
C14H15N
197.281
MFCD00059315
CWVPIIWMONJVGG-UHFFFAOYSA-N
di-m-tolylamine, 3,3'-dimethyldiphenylamine, m,m'-ditolylamine, dim-tolylamine, bis 3-methylphenyl amine, 3-methyl-n-3-methylphenyl aniline, benzenamine, 3-methyl-n-3-methylphenyl, di-m-tolyl-amine, 3,3'-ditolylamine, pubchem8963
7016139
3-methyl-N-(3-methylphenyl)aniline
CC1=CC(=CC=C1)NC2=CC=CC(=C2)C
This item is not returnable. View return policy
626-13-1
C14H15N
197.281
MFCD00059315
CWVPIIWMONJVGG-UHFFFAOYSA-N
di-m-tolylamine, 3,3'-dimethyldiphenylamine, m,m'-ditolylamine, dim-tolylamine, bis 3-methylphenyl amine, 3-methyl-n-3-methylphenyl aniline, benzenamine, 3-methyl-n-3-methylphenyl, di-m-tolyl-amine, 3,3'-ditolylamine, pubchem8963
7016139
3-methyl-N-(3-methylphenyl)aniline
CC1=CC(=CC=C1)NC2=CC=CC(=C2)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.04
Boiling Point 200°C (24 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6225
Quantity 1 g
Formula Weight 197.28
Percent Purity 98%
Chemical Name or Material 3,3'-Dimethyldiphenylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'