missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Oxycyclohexyl)indole, 95%, Thermo Scientific™

Catalog Number 15400199
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
935758-78-4
3-(3-Indolyl)cyclohexanone
This item is not returnable. View return policy
935758-78-4
3-(3-Indolyl)cyclohexanone

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 7639563
Formula Weight 213.28
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(3-Oxycyclohexyl)indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'