missing translation for 'onlineSavingsMsg'
Learn More

3',4',7,8-Tetramethoxyflavone, 97%, Thermo Scientific Chemicals

Catalog Number p-7046293
Click to view available options
Quantity:
50 mg
250 mg
This item is not returnable. View return policy
65548-55-2
C19H18O6
342.347
MFCD00143001
ZRLWYUNKZNRQLO-UHFFFAOYSA-N
3',4',7,8-tetramethoxyflavone, 2-3,4-dimethoxyphenyl-7,8-dimethoxy-4h-chromen-4-one, 2-3,4-dimethoxyphenyl-7,8-dimethoxychromen-4-one, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl-7,8-dimethoxy, 2-3,4-dimethoxy-phenyl-7,8-dimethoxy-chromen-4-one, acmc-1b8pz, 7,8,3',4'-tetramethoxyflavone, 7,8-dimethoxy-2-3,4-dimethoxyphenyl-4h-chromen-4-one
4033898
2-(3,4-dimethoxyphenyl)-7,8-dimethoxychromen-4-one
COC1=C(C=C(C=C1)C2=CC(=O)C3=C(O2)C(=C(C=C3)OC)OC)OC
This item is not returnable. View return policy
65548-55-2
C19H18O6
342.347
MFCD00143001
ZRLWYUNKZNRQLO-UHFFFAOYSA-N
3',4',7,8-tetramethoxyflavone, 2-3,4-dimethoxyphenyl-7,8-dimethoxy-4h-chromen-4-one, 2-3,4-dimethoxyphenyl-7,8-dimethoxychromen-4-one, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl-7,8-dimethoxy, 2-3,4-dimethoxy-phenyl-7,8-dimethoxy-chromen-4-one, acmc-1b8pz, 7,8,3',4'-tetramethoxyflavone, 7,8-dimethoxy-2-3,4-dimethoxyphenyl-4h-chromen-4-one
4033898
2-(3,4-dimethoxyphenyl)-7,8-dimethoxychromen-4-one
COC1=C(C=C(C=C1)C2=CC(=O)C3=C(O2)C(=C(C=C3)OC)OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 206°C to 208°C
Quantity 50 mg
Beilstein 330821
Formula Weight 342.35
Percent Purity 97%
Chemical Name or Material 3',4',7,8-Tetramethoxyflavone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'