Learn More
3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 67-03-8 |
| Molecular Formula | C12H18Cl2N4OS |
| MDL Number | MFCD00012780 |
| Quantity | 100 mg |
| SMILES | CC1=C(CCO)SC=[N+]1CC2=CN=C(C)N=C2N.[H]Cl.[Cl-] |
| Molecular Weight (g/mol) | 337.27 |
| Percent Purity | 99% |
| Chemical Name or Material | 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride |