missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenoxy)-N,N-dimethylpropylamine, 97%, Thermo Scientific™

Catalog Number p-3522473
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
76579-64-1
C11H16BrNO
258.159
MFCD03718813
CAPNMXRLKVMOOB-UHFFFAOYSA-N
3-4-bromophenoxy propyl dimethylamine, 3-4-bromophenoxy-n,n-dimethylpropylamine, 3-4-bromophenoxy-n,n-dimethylpropan-1-amine, 1-propanamine,3-4-bromophenoxy-n,n-dimethyl, 4-n,n-dimethylaminopropyloxy phenyl bromide, n,n-dimethyl-3-4-bromophenoxy-1-propanamine
2183674
3-(4-bromophenoxy)-N,N-dimethylpropan-1-amine
CN(C)CCCOC1=CC=C(C=C1)Br
This item is not returnable. View return policy
76579-64-1
C11H16BrNO
258.159
MFCD03718813
CAPNMXRLKVMOOB-UHFFFAOYSA-N
3-4-bromophenoxy propyl dimethylamine, 3-4-bromophenoxy-n,n-dimethylpropylamine, 3-4-bromophenoxy-n,n-dimethylpropan-1-amine, 1-propanamine,3-4-bromophenoxy-n,n-dimethyl, 4-n,n-dimethylaminopropyloxy phenyl bromide, n,n-dimethyl-3-4-bromophenoxy-1-propanamine
2183674
3-(4-bromophenoxy)-N,N-dimethylpropan-1-amine
CN(C)CCCOC1=CC=C(C=C1)Br
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 258.16
Percent Purity 97%
Chemical Name or Material 3-(4-Bromophenoxy)-N, N-dimethylpropylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'