Learn More
3,4-Diaminoanisole, TRC
Specifications
Specifications
| Chemical Name or Material | 3,4-Diaminoanisole |
| CAS | 102-51-2 |
| Molecular Formula | C7 H10 N2 O |
| Quantity | 500 mg |
| Synonym | 1,2-Benzenediamine, 4-methoxy-, o-Phenylenediamine, 4-methoxy- (6CI,7CI,8CI), 4-Methoxy-1,2-benzenediamine, 1,2-Diamino-4-methoxybenzene, 2-Amino-4-methoxy aniline, 3,4-Diaminoanisole, 4-Methoxy-1,2-diaminobenzene, 4-Methoxy-1,2-phenylenediamine, 4-Methoxy-2-aminoaniline, 4-Methoxy-o-phenylenediamine, p-Methoxy-o-phenylenediamine |
| InChI Formula | InChI=1S/C7H10N2O/c1-10-5-2-3-6(8)7(9)4-5/h2-4H,8-9H2,1H3 |
| SMILES | COc1ccc(N)c(N)c1 |
| IUPAC Name | 4-methoxybenzene-1,2-diamine |
| Molecular Weight (g/mol) | 138.17 |
| Formula Weight | 138.0793 |