Learn More
3',4'-Dichloropropionanilide-2,2,3,3,3-d5, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Propanil-d5 |
| CAS | 1398065-82-1 |
| Molecular Formula | C9 D5 H4 Cl2 N O |
| Quantity | 0.05 g |
| Synonym | Propanil-d5, Propanil-D5 (propanoyl-D5), 3',4'-Dichloropropionanilide-2,2,3,3,3-d5, N-(3,4-Dichlorophenyl)propanamide-2,2,3,3,3-d5, N-(3,4-Dichlorophenyl)propionamide-2,2,3,3,3-d5 |
| InChI Formula | InChI=1 S/C9H9Cl2NO/c1-2-9(13)12-6-3-4-7(10)8(11)5-6/h3-5 H,2H2,1H3,(H,12,13)/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C(=O)Nc1ccc(Cl)c(Cl)c1 |
| IUPAC Name | 2,2,3,3,3-pentadeuterio-N-(3,4-dichlorophenyl)propanamide |
| Molecular Weight (g/mol) | 223.111 |
| Formula Weight | 222.038 g/mol |
| Show More |