missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzamidoxime, 97%, Thermo Scientific™

Product Code. 15490868
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490868 250 mg 250mg
15470868 1 g 1g
15480868 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490868 Supplier Thermo Scientific Alfa Aesar Supplier No. H54636.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500024-74-8
Molecular Formula C7H6F2N2O
Molecular Weight (g/mol) 172.14
MDL Number MFCD06660194,MFCD06796653
InChI Key OGCLGUHODKKLSH-UHFFFAOYSA-N
Synonym 3,4-difluorobenzamidoxime, 3,4-difluoro-n-hydroxy-benzamidine, 3,4-difluoro-n'-hydroxybenzene-1-carboximidamide, 3,4-difluorophenyl hydroxyimino methylamine, benzenecarboximidamide, 3,4-difluoro-n-hydroxy, z-3,4-difluoro-n'-hydroxybenzene-1-carboximidamide, e-3,4-difluoro-n'-hydroxybenzenecarboximidamide
PubChem CID 17750753
IUPAC Name 3,4-difluoro-N'-hydroxybenzenecarboximidamide
SMILES NC(=NO)C1=CC(F)=C(F)C=C1

Specifications

Quantity 250 mg
Beilstein 19347401
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material 3,4-Difluorobenzamidoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.