Learn More
(-)-3,4-Dihydroxy-alpha-methyl-L-phenylalanine sesquihydrate, 99%
Product Code. 15432846
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15432846 5 g 5g
15422846 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15432846 Supplier Thermo Scientific Alfa Aesar Supplier No. H56704.06
Request Bulk or Custom Format
CAS: 41372-08-1 | C10H15NO5 | 229.232 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 41372-08-1
Molecular Formula C10H15NO5
Molecular Weight (g/mol) 229.232
MDL Number MFCD00149280
InChI Key XDJUBZFLFDODNF-PPHPATTJSA-N
Synonym methyldopa hydrate, l-methyldopa, s-2-amino-3-3,4-dihydroxyphenyl-2-methylpropionic acid sesquihydrate, --3-3,4-dihydroxyphenyl-2-methyl-l-alanine sesquihydrate, l---alpha-methyldopa hydrate, alpha-methyl-l-dopa sesquihydrate, --3,4-dihydroxy-^a-methyl-l-phenylalanine sesquihydrate, --3-3,4-dihydroxyphenyl-2-methyl-l-alanine ses, 2s-2-amino-3-3,4-dihydroxyphenyl-2-methylpropanoic acid, oxamethane
PubChem CID 18530292
IUPAC Name (2S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoic acid;hydrate
SMILES CC(CC1=CC(=C(C=C1)O)O)(C(=O)O)N.O
Specifications
Melting Point >300°C
Merck Index 14,6055
Quantity 5 g
Formula Weight 238.24 (211.22 Anhydrous)
Optical Rotation −13° (c=1 in Water)
Sensitivity Air and light sensitive
Percent Purity 99%
Chemical Name or Material (-)-3,4-Dihydroxy-alpha-methyl-L-phenylalanine sesquihydrate