missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenylacetone, 97%, Thermo Scientific™

Catalog Number p-7077171
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
776-99-8
C11H14O3
194.23
MFCD00008772
UMYZWICEDUEWIM-UHFFFAOYSA-N
3,4-dimethoxyphenylacetone, 1-3,4-dimethoxyphenyl propan-2-one, 3,4-dimethoxyphenyl acetone, veratryl-2-propanone, 1-3,4-dimethoxyphenyl-2-propanone, veratryl acetone, 3,4-dimethoxybenzyl methyl ketone, 1-3,4-dimethoxyphenyl acetone, 2-propanone, 1-3,4-dimethoxyphenyl, veratryl methyl ketone
69896
1-(3,4-dimethoxyphenyl)propan-2-one
COC1=CC=C(CC(C)=O)C=C1OC
This item is not returnable. View return policy
776-99-8
C11H14O3
194.23
MFCD00008772
UMYZWICEDUEWIM-UHFFFAOYSA-N
3,4-dimethoxyphenylacetone, 1-3,4-dimethoxyphenyl propan-2-one, 3,4-dimethoxyphenyl acetone, veratryl-2-propanone, 1-3,4-dimethoxyphenyl-2-propanone, veratryl acetone, 3,4-dimethoxybenzyl methyl ketone, 1-3,4-dimethoxyphenyl acetone, 2-propanone, 1-3,4-dimethoxyphenyl, veratryl methyl ketone
69896
1-(3,4-dimethoxyphenyl)propan-2-one
COC1=CC=C(CC(C)=O)C=C1OC

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.1150g/mL
Boiling Point 154.0°C to 158.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula (CH3O)2C6H3CH2COCH3
Refractive Index 1.5338 to 1.5358
Quantity 5 g
Specific Gravity 1.115
Formula Weight 194.23
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxyphenylacetone
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'