missing translation for 'onlineSavingsMsg'
Learn More

3,4-Ethylenedioxythiophene, 99%, Thermo Scientific Chemicals

Catalog Number p-7077522
Click to view available options
Quantity:
5 g
25 g
126213-50-1
C6H6O2S
142.18
GKWLILHTTGWKLQ-UHFFFAOYSA-N
3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, 2h,3h-thiopheno 3,4-e 1,4-dioxane
4421864
2,3-dihydrothieno[3,4-b][1,4]dioxine
C1COC2=CSC=C2O1
126213-50-1
C6H6O2S
142.18
GKWLILHTTGWKLQ-UHFFFAOYSA-N
3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, 2h,3h-thiopheno 3,4-e 1,4-dioxane
4421864
2,3-dihydrothieno[3,4-b][1,4]dioxine
C1COC2=CSC=C2O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 10.5°C to 12.0°C
Density 1.3310g/mL
Boiling Point 193.0°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Quantity 5 g
Specific Gravity 1.331
Formula Weight 142.18
Percent Purity 99%
Chemical Name or Material 3,4-Ethylenedioxythiophene
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Ethylenedioxythiophene, 99%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'