Learn More
3-(4-Fluorophenyl)-1H-indole, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(4-Fluorophenyl)-1H-indole |
| CAS | 101125-32-0 |
| Molecular Formula | C14H10FN |
| Quantity | 2.5 g |
| Synonym | 3-(4-Fluorophenyl)indole |
| InChI Formula | InChI=1S/C14H10FN/c15-11-7-5-10(6-8-11)13-9-16-14-4-2-1-3-12(13)14/h1-9,16H |
| SMILES | Fc1ccc(cc1)c2c[nH]c3ccccc23 |
| IUPAC Name | 3-(4-fluorophenyl)-1H-indole |
| Molecular Weight (g/mol) | 211.23 |
| Formula Weight | 211.0797 |