missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methoxybenzylideneamino)benzeneboronic acid pinacol ester, Thermo Scientific™

Product Code. 15499175
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499175 5 g 5g
15489175 1 g 1g
15409185 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15499175 Supplier Thermo Scientific Alfa Aesar Supplier No. H51129.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380151-91-7
Molecular Formula C20H24BNO3
Molecular Weight (g/mol) 337.226
MDL Number MFCD11113043
InChI Key OKCWNDNFJLMDKV-UHFFFAOYSA-N
Synonym e-n-4-methoxybenzylidene-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-4-methoxybenzylidineamino benzeneboronic acid pinacol ester, 3-4-methoxybenzylidine-3-aminophenylboronic acid pinacol ester, 1e-1-4-methoxyphenyl-n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanimine, amtb127, 3-4-methoxybenzylideneamino benzeneboronic acid pinacol ester, ne-n-4-methoxyphenyl methylidene-3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 1-4-methoxyphenyl-n-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanimine, 2-3-4-methoxybenzylidene amino phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, n-4-methoxybenzylidene-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline
PubChem CID 11078232
IUPAC Name 1-(4-methoxyphenyl)-N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanimine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N=CC3=CC=C(C=C3)OC

Specifications

Quantity 5 g
Formula Weight 337.22
Chemical Name or Material 3-(4-Methoxybenzylideneamino)benzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.