Learn More
3-[(4-Methylperhydro-1,4-diazepin-1-yl)methyl]benzonitrile, 97%, Thermo Scientific™
Product Code. 10402854
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10402854 1 g Amber Glass Bottle 1g
11396795 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10402854 Supplier Thermo Scientific Maybridge Supplier No. CC55916DA
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Chemical Identifiers
CAS 910036-91-8
Molecular Formula C14H19N3
Molecular Weight (g/mol) 229.327
MDL Number MFCD09702379
InChI Key WCQJGGDNUFLTPA-UHFFFAOYSA-N
Synonym 3-4-methylhomopiperazin-1-yl methyl benzonitrile, 3-4-methyl-1,4-diazepan-1-yl methyl benzonitrile, 3-4-methylperhydro-1,4-diazepin-1-yl methyl benzonitrile, 3-4-methyl-1,4-diazaperhydroepinyl methyl benzenecarbonitrile, benzonitrile,3-hexahydro-4-methyl-1h-1,4-diazepin-1-yl methyl
PubChem CID 24229556
IUPAC Name 3-[(4-methyl-1,4-diazepan-1-yl)methyl]benzonitrile
SMILES CN1CCCN(CC1)CC2=CC=CC(=C2)C#N
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 229.33
Percent Purity 97%
Chemical Name or Material 3-[(4-Methylperhydro-1, 4-diazepin-1-yl)methyl]benzonitrile