missing translation for 'onlineSavingsMsg'
Learn More

(3-(4-(Pyrimidin-2-yl)piperazin-1-yl)azetidin-1-yl)(1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1H-indol-5-yl)methanone, TRC

Product Code. 30472006
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472006 1 mg 1mg
30483897 5 mg 5mg
30475498 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30472006 Supplier Toronto Research Chemicals Supplier No. TRCM5581651MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3-(4-(Pyrimidin-2-yl)piperazin-1-yl)azetidin-1-yl)(1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1H-indol-5-yl)methanone
Analyte or Component Names (3-(4-(Pyrimidin-2-yl)piperazin-1-yl)azetidin-1-yl)(1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1H-indol-5-yl)methanone
Shipping Condition Room Temperature
Molecular Formula C11 H18 N5 O13 P3 S . C6 H15 N
Synonym MAGL-2102-precursor
InChI Formula InChI=1S/C11H18N5O13P3S.C6H15N/c1-33-9-5-8(14-11(12)15-9)16(3-13-5)10-7(18)6(17)4(27-10)2-26-31(22,23)29-32(24,25)28-30(19,20)21;1-4-7(5-2)6-3/h3-4,6-7,10,17-18H,2H2,1H3,(H,22,23)(H,24,25)(H2,12,14,15)(H2,19,20,21);4-6H2,1-3H3/t4-,6-,7-,10-;/m1./s1
Solution Type Neat
SMILES CCN(CC)CC.CSc1nc(N)nc2c1ncn2[C@@H]3O[C@H](COP(=O)(O)OP(=O)(O)OP(=O)(O)O)[C@@H](O)[C@H]3O
IUPAC Name [[(2R,3S,4R,5R)-5-(2-amino-6-methylsulfanylpurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;N,N-diethylethanamine
Molecular Weight (g/mol) 654.463
Quantity 1 mg
Formula Weight 654.104
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.