Learn More
3-[5-(2-thienyl)-2-furyl]propanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-[5-(2-thienyl)-2-furyl]propanoic Acid |
| CAS | 24090-38-8 |
| Molecular Formula | C11 H10 O3 S |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C11H10O3S/c12-11(13)6-4-8-3-5-9(14-8)10-2-1-7-15-10/h1-3,5,7H,4,6H2,(H,12,13) |
| SMILES | OC(=O)CCc1oc(cc1)c2cccs2 |
| IUPAC Name | 3-(5-thiophen-2-ylfuran-2-yl)propanoic acid |
| Molecular Weight (g/mol) | 222.26 |
| Formula Weight | 222.035 |