Learn More
3,5,6-Trichloro-8-quinolinecarboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3,5,6-Trichloro-8-quinolinecarboxylic Acid |
| CAS | 1391053-30-7 |
| Molecular Formula | C10H4Cl3NO2 |
| Quantity | 5 mg |
| Synonym | 3,5,6-Trichloroquinoline-8-carboxylic Acid |
| InChI Formula | InChI=1S/C10H4Cl3NO2/c11-4-1-5-8(13)7(12)2-6(10(15)16)9(5)14-3-4/h1-3H,(H,15,16) |
| SMILES | OC(=O)c1cc(Cl)c(Cl)c2cc(Cl)cnc12 |
| IUPAC Name | 3,5,6-trichloroquinoline-8-carboxylic acid |
| Molecular Weight (g/mol) | 276.5 |
| Formula Weight | 274.9308 |